missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl Isopropylmalonate, TCI America™

$62.64 - $508.77

Chemical Identifiers

CAS 759-36-4
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00040491
InChI Key BYQFBFWERHXONI-UHFFFAOYSA-N
Synonym diethyl isopropylmalonate, diethyl 2-isopropylmalonate, isopropyl diethyl malonate, 1,3-diethyl 2-propan-2-yl propanedioate, diethylisopropylmalonate, isopropylmalonic acid diethyl ester, propanedioic acid, 1-methylethyl-, diethyl ester, ethyl isopropylmalonate, 1,3-diethyl 2-isopropylpropanedioate, 2-isopropyl-malonic acid diethyl ester
PubChem CID 12966
IUPAC Name diethyl 2-propan-2-ylpropanedioate
SMILES CCOC(=O)C(C(C)C)C(=O)OCC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I0252500ML View Documents TCI America
I0252500ML
500mL Each for $508.77
 
I0252100ML View Documents TCI America
I0252100ML
100mL Each for $155.84
 
I025225ML View Documents TCI America
I025225ML
25mL Each for $62.64
 
Specifications

Chemical Identifiers

759-36-4
202.25
BYQFBFWERHXONI-UHFFFAOYSA-N
12966
CCOC(=O)C(C(C)C)C(=O)OCC
C10H18O4
MFCD00040491
diethyl isopropylmalonate, diethyl 2-isopropylmalonate, isopropyl diethyl malonate, 1,3-diethyl 2-propan-2-yl propanedioate, diethylisopropylmalonate, isopropylmalonic acid diethyl ester, propanedioic acid, 1-methylethyl-, diethyl ester, ethyl isopropylmalonate, 1,3-diethyl 2-isopropylpropanedioate, 2-isopropyl-malonic acid diethyl ester
diethyl 2-propan-2-ylpropanedioate

Specifications

214°C
500mL
C10H18O4
diethyl isopropylmalonate, diethyl 2-isopropylmalonate, isopropyl diethyl malonate, 1,3-diethyl 2-propan-2-yl propanedioate, diethylisopropylmalonate, isopropylmalonic acid diethyl ester, propanedioic acid, 1-methylethyl-, diethyl ester, ethyl isopropylmalonate, 1,3-diethyl 2-isopropylpropanedioate, 2-isopropyl-malonic acid diethyl ester
CCOC(=O)C(C(C)C)C(=O)OCC
202.25
202.25
≥98.0% (GC)
Yellow
759-36-4
MFCD00040491
BYQFBFWERHXONI-UHFFFAOYSA-N
diethyl 2-propan-2-ylpropanedioate
12966
Clear Liquid at 20°C
Diethyl Isopropylmalonate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0879

TSCA : No