missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl Isopropylidenesuccinate, TCI America™

$163.47 - $484.31

Chemical Identifiers

CAS 42103-98-0
Molecular Formula C11H18O4
Molecular Weight (g/mol) 214.261
MDL Number MFCD00130132
InChI Key FSUBDXPWXDGTBG-UHFFFAOYSA-N
Synonym Isopropylidenesuccinic Acid Diethyl Ester
PubChem CID 251287
IUPAC Name diethyl 2-propan-2-ylidenebutanedioate
SMILES CCOC(=O)CC(=C(C)C)C(=O)OCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I04045G View Documents TCI America
I04045G
5g Each for $484.31
 
I04041G View Documents TCI America
I04041G
1g Each for $163.47
 
Specifications

Chemical Identifiers

42103-98-0
214.261
FSUBDXPWXDGTBG-UHFFFAOYSA-N
251287
CCOC(=O)CC(=C(C)C)C(=O)OCC
C11H18O4
MFCD00130132
Isopropylidenesuccinic Acid Diethyl Ester
diethyl 2-propan-2-ylidenebutanedioate

Specifications

103°C
5g
C11H18O4
Isopropylidenesuccinic Acid Diethyl Ester
CCOC(=O)CC(=C(C)C)C(=O)OCC
214.261
214.26
≥98.0% (GC)
Colorless
42103-98-0
MFCD00130132
FSUBDXPWXDGTBG-UHFFFAOYSA-N
diethyl 2-propan-2-ylidenebutanedioate
251287
Clear Liquid at 20°C
Diethyl Isopropylidenesuccinate
Safety and Handling

Safety and Handling

TSCA : No