Please login to your online account to display your discounted pricing

Diethyl isopropylidenemalonate, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material Diethyl isopropylidenemalonate
Assay Percent Range 97%
Molecular Formula C10H16O4
Formula Weight 200.23
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC443310250

 
thermo fisher scientific
AC443310250
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC443310050

 
thermo fisher scientific
AC443310050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Diethyl isopropylidenemalonate
Quantity 25g
CAS 6802-75-1
Assay Percent Range 97%
Molecular Formula C10H16O4
Formula Weight 200.23