missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diethyl isopropylidenemalonate, 97%, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific AC443310250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Diethyl isopropylidenemalonate | |
1.0210g/mL | |
96% min. (GC) | |
1,3-diethyl 2-propan-2-ylidene propanedioate, 2-isopropylidene-malonic acid diethyl ester, acmc-209o1m, diethyl 2-1-methylethylidene malonate, diethyl 2-propan-2-ylidene malonate, diethyl isopropylidenemalonate, diethylisopropylidenemalonate, isopropylidenemalonic acid diethyl ester, malonic acid, isopropylidene-, diethyl ester, propanedioic acid, 1-methylethylidene-, diethyl ester | |
WEISAZNMMVPNTH-UHFFFAOYSA-N | |
CCOC(=O)C(=C(C)C)C(=O)OCC | |
200.23 | |
200.23 |
6802-75-1 | |
175°C to 178°C (120.0mmHg) | |
C10H16O4 | |
25g | |
1.021 | |
diethyl 2-propan-2-ylidenepropanedioate | |
81255 | |
97% |
Chemical Identifiers
6802-75-1 | |
200.23 | |
1,3-diethyl 2-propan-2-ylidene propanedioate, 2-isopropylidene-malonic acid diethyl ester, acmc-209o1m, diethyl 2-1-methylethylidene malonate, diethyl 2-propan-2-ylidene malonate, diethyl isopropylidenemalonate, diethylisopropylidenemalonate, isopropylidenemalonic acid diethyl ester, malonic acid, isopropylidene-, diethyl ester, propanedioic acid, 1-methylethylidene-, diethyl ester | |
diethyl 2-propan-2-ylidenepropanedioate |
C10H16O4 | |
WEISAZNMMVPNTH-UHFFFAOYSA-N | |
81255 | |
CCOC(=O)C(=C(C)C)C(=O)OCC |
Safety and Handling
EINECSNumber : 229-876-4
RUO – Research Use Only