missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl isopropylidenemalonate, 97%

Manufacturer:  Alfa Aesar™ L1119514

 View more versions of this product

Catalog No. AAL1119514



Specifications

Specifications

Diethyl isopropylidenemalonate
123°C to 126°C (12mmHg)
6802-75-1
MFCD00009147
diethyl isopropylidenemalonate, diethyl 2-propan-2-ylidene malonate, diethylisopropylidenemalonate, malonic acid, isopropylidene-, diethyl ester, propanedioic acid, 1-methylethylidene-, diethyl ester, isopropylidenemalonic acid diethyl ester, 1,3-diethyl 2-propan-2-ylidene propanedioate, diethyl 2-1-methylethylidene malonate, 2-isopropylidene-malonic acid diethyl ester, acmc-209o1m
CCOC(=O)C(=C(C)C)C(=O)OCC
200.234
200.24
109°C (228°F)
25g
C10H16O4
1776122
WEISAZNMMVPNTH-UHFFFAOYSA-N
diethyl 2-propan-2-ylidenepropanedioate
81255
97%

Chemical Identifiers

6802-75-1
200.234
WEISAZNMMVPNTH-UHFFFAOYSA-N
81255
CCOC(=O)C(=C(C)C)C(=O)OCC
C10H16O4
MFCD00009147
diethyl isopropylidenemalonate, diethyl 2-propan-2-ylidene malonate, diethylisopropylidenemalonate, malonic acid, isopropylidene-, diethyl ester, propanedioic acid, 1-methylethylidene-, diethyl ester, isopropylidenemalonic acid diethyl ester, 1,3-diethyl 2-propan-2-ylidene propanedioate, diethyl 2-1-methylethylidene malonate, 2-isopropylidene-malonic acid diethyl ester, acmc-209o1m
diethyl 2-propan-2-ylidenepropanedioate
Documents