missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diethyl isopropylidenemalonate, 97%, Thermo Scientific Chemicals
$61.92 - $61.92
Chemical Identifiers
CAS | 6802-75-1 |
---|---|
Molecular Formula | C10H16O4 |
Molecular Weight (g/mol) | 200.234 |
MDL Number | MFCD00009147 |
InChI Key | WEISAZNMMVPNTH-UHFFFAOYSA-N |
Synonym | diethyl isopropylidenemalonate, diethyl 2-propan-2-ylidene malonate, diethylisopropylidenemalonate, malonic acid, isopropylidene-, diethyl ester, propanedioic acid, 1-methylethylidene-, diethyl ester, isopropylidenemalonic acid diethyl ester, 1,3-diethyl 2-propan-2-ylidene propanedioate, diethyl 2-1-methylethylidene malonate, 2-isopropylidene-malonic acid diethyl ester, acmc-209o1m |
PubChem CID | 81255 |
IUPAC Name | diethyl 2-propan-2-ylidenepropanedioate |
SMILES | CCOC(=O)C(=C(C)C)C(=O)OCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1119506
|
Thermo Scientific Chemicals
L1119506 |
5 g |
Each for $61.92
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6802-75-1 | |
200.234 | |
WEISAZNMMVPNTH-UHFFFAOYSA-N | |
81255 | |
CCOC(=O)C(=C(C)C)C(=O)OCC |
C10H16O4 | |
MFCD00009147 | |
diethyl isopropylidenemalonate, diethyl 2-propan-2-ylidene malonate, diethylisopropylidenemalonate, malonic acid, isopropylidene-, diethyl ester, propanedioic acid, 1-methylethylidene-, diethyl ester, isopropylidenemalonic acid diethyl ester, 1,3-diethyl 2-propan-2-ylidene propanedioate, diethyl 2-1-methylethylidene malonate, 2-isopropylidene-malonic acid diethyl ester, acmc-209o1m | |
diethyl 2-propan-2-ylidenepropanedioate |
Specifications
6802-75-1 | |
123°C to 126°C (12 mmHg) | |
C10H16O4 | |
MFCD00009147 | |
1776122 | |
WEISAZNMMVPNTH-UHFFFAOYSA-N | |
diethyl 2-propan-2-ylidenepropanedioate | |
81255 | |
97% |
1.021 | |
109°C (228°F) | |
1.45 | |
5 g | |
diethyl isopropylidenemalonate, diethyl 2-propan-2-ylidene malonate, diethylisopropylidenemalonate, malonic acid, isopropylidene-, diethyl ester, propanedioic acid, 1-methylethylidene-, diethyl ester, isopropylidenemalonic acid diethyl ester, 1,3-diethyl 2-propan-2-ylidene propanedioate, diethyl 2-1-methylethylidene malonate, 2-isopropylidene-malonic acid diethyl ester, acmc-209o1m | |
CCOC(=O)C(=C(C)C)C(=O)OCC | |
200.234 | |
200.24 | |
Diethyl isopropylidenemalonate |
Safety and Handling
EINECSNumber : 229-876-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only