missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl indole-2,6-dicarboxylate, 96%

Manufacturer:  Alfa Aesar™ H54619MD

 View more versions of this product

Catalog No. AAH54619MD



Specifications

Specifications

Diethyl indole-2,6-dicarboxylate
250mg
C14H15NO4
1h-indole-2,6-dicarboxylic acid diethyl ester
CCOC(=O)C1=CC2=C(C=C1)C=C(N2)C(=O)OCC
261.277
261.27
119°C to 121°C
107516-75-6
MFCD03411566
ISQYBMUDGXFHAF-UHFFFAOYSA-N
diethyl 1H-indole-2,6-dicarboxylate
44754779
96%

Chemical Identifiers

107516-75-6
261.277
ISQYBMUDGXFHAF-UHFFFAOYSA-N
44754779
CCOC(=O)C1=CC2=C(C=C1)C=C(N2)C(=O)OCC
C14H15NO4
MFCD03411566
1h-indole-2,6-dicarboxylic acid diethyl ester
diethyl 1H-indole-2,6-dicarboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents