missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl Iminodiacetate, TCI America™

$44.31 - $128.34

Chemical Identifiers

CAS 6290-05-7
Molecular Formula C8H15NO4
Molecular Weight (g/mol) 189.21
MDL Number MFCD00041925
InChI Key LJDNMOCAQVXVKY-UHFFFAOYSA-N
Synonym diethyl iminodiacetate, diethyl 2,2'-azanediyldiacetate, diethyliminodiacetate, iminodiacetic acid diethyl ester, glycine, n-2-ethoxy-2-oxoethyl-, ethyl ester, diethyl 2,2'-iminodiacetate, n,n-bis ethoxycarbonylmethyl amine, acetic acid, iminodi-, diethyl ester, ethyl 2-2-ethoxy-2-oxoethyl amino acetate, zlchem 799
PubChem CID 80502
IUPAC Name ethyl 2-[(2-ethoxy-2-oxoethyl)amino]acetate
SMILES CCOC(=O)CNCC(=O)OCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I03265ML View Documents TCI America
I03265ML
5mL Each for $44.31
 
I032625ML View Documents TCI America
I032625ML
25mL Each for $128.34
 
Specifications

Chemical Identifiers

6290-05-7
189.21
LJDNMOCAQVXVKY-UHFFFAOYSA-N
80502
CCOC(=O)CNCC(=O)OCC
C8H15NO4
MFCD00041925
diethyl iminodiacetate, diethyl 2,2'-azanediyldiacetate, diethyliminodiacetate, iminodiacetic acid diethyl ester, glycine, n-2-ethoxy-2-oxoethyl-, ethyl ester, diethyl 2,2'-iminodiacetate, n,n-bis ethoxycarbonylmethyl amine, acetic acid, iminodi-, diethyl ester, ethyl 2-2-ethoxy-2-oxoethyl amino acetate, zlchem 799
ethyl 2-[(2-ethoxy-2-oxoethyl)amino]acetate

Specifications

122°C
5mL
C8H15NO4
diethyl iminodiacetate, diethyl 2,2'-azanediyldiacetate, diethyliminodiacetate, iminodiacetic acid diethyl ester, glycine, n-2-ethoxy-2-oxoethyl-, ethyl ester, diethyl 2,2'-iminodiacetate, n,n-bis ethoxycarbonylmethyl amine, acetic acid, iminodi-, diethyl ester, ethyl 2-2-ethoxy-2-oxoethyl amino acetate, zlchem 799
CCOC(=O)CNCC(=O)OCC
189.21
189.21
≥98.0% (GC)
Yellow
6290-05-7
MFCD00041925
LJDNMOCAQVXVKY-UHFFFAOYSA-N
ethyl 2-[(2-ethoxy-2-oxoethyl)amino]acetate
80502
Clear Liquid at 20°C
Diethyl Iminodiacetate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3117

TSCA : Yes