Please login to your online account to display your discounted pricing

Diethyl iminodiacetate, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Diethyl iminodiacetate
Name Note 99%
CAS 1603543
Assay ≥97.5% (GC)
Assay Percent Range 99%
Molecular Formula C8H15NO4
Linear Formula HN(CH2COOC2H5)2
Formula Weight 189.21
MDL Number MFCD00041925
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC407320050

  SDS
thermo fisher scientific
AC407320050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC407320250

  SDS
fisher scientific acros
407320250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC407321000

  SDS
fisher scientific acros
407321000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5% (GC)
Physical Form Viscous Liquid
Formula Weight 189.21
Quantity 5g
Color Clear light yellow to yellow
TSCA TSCA
EINECS Number 228-533-6
Density 1.0800g/mL
CAS 1603543
Refractive Index 1.4345 to 1.4385 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula HN(CH2COOC2H5)2
Chemical Name or Material Diethyl iminodiacetate
Name Note 99%
Assay Percent Range 99%
Molecular Formula C8H15NO4
MDL Number MFCD00041925
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.