missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl iminodiacetate, 97%

$41.32 - $144.96

Chemical Identifiers

CAS 6299-02-1
Molecular Formula C8H10ClN
Molecular Weight (g/mol) 155.63
MDL Number MFCD00041925
InChI Key PINPOEWMCLFRRB-UHFFFAOYNA-N
Synonym 1-4-chlorophenyl ethanamine, 1-4-chlorophenyl ethylamine, 4-chloro-alpha-methylbenzylamine, 1-4-chlorophenyl ethan-1-amine, 4-chloro-alpha-methyl benzylamine, 1-4-chlorophenyl-1-ethanamine, 1-4-chloro-phenyl-ethylamine, acmc-20amkz, acmc-20apje, acmc-209jma
PubChem CID 97803
IUPAC Name 1-(4-chlorophenyl)ethanamine
SMILES CC(C1=CC=C(C=C1)Cl)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60608AA View Documents Alfa Aesar™
H60608AA
5mL Each for $41.32
 
AAH60608AC View Documents Alfa Aesar™
H60608AC
25mL Each for $144.96
 
Description

Description

Applications
It is mainly used for the manufacture of N-(Phosphonomethyl) iminodiacetic acid and glyphosate. Used as raw material for ion exchange resin; for rubbers, electroplating and food additives.

Solubility
Moderately soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications

Chemical Identifiers

6299-02-1
155.63
PINPOEWMCLFRRB-UHFFFAOYNA-N
97803
CC(C1=CC=C(C=C1)Cl)N
C8H10ClN
MFCD00041925
1-4-chlorophenyl ethanamine, 1-4-chlorophenyl ethylamine, 4-chloro-alpha-methylbenzylamine, 1-4-chlorophenyl ethan-1-amine, 4-chloro-alpha-methyl benzylamine, 1-4-chlorophenyl-1-ethanamine, 1-4-chloro-phenyl-ethylamine, acmc-20amkz, acmc-20apje, acmc-209jma
1-(4-chlorophenyl)ethanamine

Specifications

113°C (235°F)
5mL
C8H10ClN
MFCD00041925
PINPOEWMCLFRRB-UHFFFAOYNA-N
1-(4-chlorophenyl)ethanamine
97803
97%
208°C
6299-02-1
HN(CH2CO2C2H5)2
1-4-chlorophenyl ethanamine, 1-4-chlorophenyl ethylamine, 4-chloro-alpha-methylbenzylamine, 1-4-chlorophenyl ethan-1-amine, 4-chloro-alpha-methyl benzylamine, 1-4-chlorophenyl-1-ethanamine, 1-4-chloro-phenyl-ethylamine, acmc-20amkz, acmc-20apje, acmc-209jma
CC(C1=CC=C(C=C1)Cl)N
155.63
189.21
Diethyl iminodiacetate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA