Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl hydroxymethylphosphonate, tech. 85%

$52.88 - $142.84

Chemical Identifiers

Chemical Name or Material Diethyl hydroxymethylphosphonate
CAS 3084-40-0
Purity Grade Technical Grade
Molecular Formula C5H13O4P
Formula Weight 168.13
MDL Number MFCD00014418
Synonym Hydroxymethylphosphonic acid diethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2121506

  SDS
alfa aesar™
B2121506
5g Each for $52.88

AAB2121514

  SDS
alfa aesar™
B2121514
25g Each for $142.84
Description & Specifications

Specifications

Formula Weight 168.13
Synonym Hydroxymethylphosphonic acid diethyl ester
Purity Grade Technical Grade
Quantity 5g
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C5H13O4P
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 85%
Flash Point 110°C (230°F)
Chemical Name or Material Diethyl hydroxymethylphosphonate
TSCA TSCA
MDL Number MFCD00014418
CAS 3084-40-0
Boiling Point 124° to 126°C (3mmHg)

Applications
Diethyl hydroxymethylphosphonate acts as a reactant for synthesis of α And β-dialkoxyphosphoryl isothiocyanate used in antiproliferative studies, dendritic polyglycerol anions for L-selectin inhibition, phosphonates for anti-HIV-1 activity, homo and copolymers of phosphonated norbornenes, Mitsunobu reactions. It is involved in studies of FBPase inhibition by phosphonic acid-containing thaizoles.

Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in a well sealed container.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.