Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl hexafluoroglutarate, 97%

$206.43 - $785.03

Chemical Identifiers

Chemical Name or Material Diethyl hexafluoroglutarate
CAS 424-40-8
Molecular Formula C9H10F6O4
Formula Weight 296.16
MDL Number MFCD00015156
Beilstein 1804802
Synonym Diethyl perfluoroglutarate; Hexafluoroglutaric acid diethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2242806

  SDS
alfa aesar™
B2242806
5g Each for $206.43

AAB2242814

  SDS
alfa aesar™
B2242814
25g Each for $785.03
Description & Specifications

Specifications

Formula Weight 296.16
TSCA TSCA
Chemical Name or Material Diethyl hexafluoroglutarate
Beilstein 1804802
Assay 97%
Quantity 5g
Molecular Formula C9H10F6O4
Health Hazard 1 Warning
RTECS Number MA3750500
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
MDL Number MFCD00015156
Synonym Diethyl perfluoroglutarate; Hexafluoroglutaric acid diethyl ester
Health Hazard 2 GHS H Statement
H332-H315-H319-H335-H227
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
CAS 424-40-8
Boiling Point 224°C
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H332-H315-H319-H335-H227
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.