Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl glutarate, 98%

$54.19 - $134.69

Chemical Identifiers

Chemical Name or Material Diethyl glutarate
CAS 818-38-2
Molecular Formula C9H16O4
Formula Weight 188.23
MDL Number MFCD00009212
Beilstein 1777327
Synonym Glutaric acid diethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1125914

 
alfa aesar™
L1125914
25g Each for $54.19

AAL1125922

 
alfa aesar™
L1125922
100g Each for $134.69
Description & Specifications

Specifications

Formula Weight 188.23
Chemical Name or Material Diethyl glutarate
Flash Point 96°C (204°F)
Molecular Formula C9H16O4
MDL Number MFCD00009212
CAS 818-38-2
TSCA TSCA
Assay 98%
Beilstein 1777327
Synonym Glutaric acid diethyl ester
Boiling Point 236° to 237°C
Quantity 25g