Learn More
Diethyl fumarate, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC114380025
Specifications
Diethyl fumarate | |
91°C | |
Glass bottle | |
1.05 | |
Colorless to Yellow | |
2.5kg | |
623-91-6 | |
C8H12O4 | |
MFCD00064455 | |
2-butenedioic acid 2e-, diethyl ester, 2-butenedioic acid e-, diethyl ester, anti-psoriaticum, diethyl 2e-but-2-enedioate, diethyl e-but-2-enedioate, diethyl fumarate, fumaric acid diethyl ester, fumaric acid, diethyl ester, trans-2-butenedioic acid diethyl ester, unii-5wbu5a3e8a | |
IEPRKVQEAMIZSS-AATRIKPKSA-N | |
diethyl (E)-but-2-enedioate | |
638144 | |
172.18 | |
98% |
1.0500g/mL | |
Authentic | |
1.4396 to 1.4416 | |
218.0°C to 219.0°C | |
1.0°C to 2.0°C | |
98% | |
98% | |
C2H5O2CCH=CHCO2C2H5 | |
02,742 | |
Solubility in water: immiscible. | |
CCOC(=O)C=CC(=O)OCC | |
172.18 | |
CHEBI:87388 | |
Liquid |
Chemical Identifiers
623-91-6 | |
172.18 | |
IEPRKVQEAMIZSS-AATRIKPKSA-N | |
638144 | |
diethyl (E)-but-2-enedioate |
C8H12O4 | |
MFCD00064455 | |
2-butenedioic acid 2e-, diethyl ester, 2-butenedioic acid e-, diethyl ester, anti-psoriaticum, diethyl 2e-but-2-enedioate, diethyl e-but-2-enedioate, diethyl fumarate, fumaric acid diethyl ester, fumaric acid, diethyl ester, trans-2-butenedioic acid diethyl ester, unii-5wbu5a3e8a | |
CHEBI:87388 | |
CCOC(=O)C=CC(=O)OCC |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 210-819-7
RTECSNumber : EM5950000
TSCA : TSCA