Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl fumarate, 98%

$43.41 - $538.74

Chemical Identifiers

Chemical Name or Material Diethyl fumarate
CAS 623-91-6
Molecular Formula C8H12O4
Formula Weight 172.18
MDL Number MFCD00064455
Beilstein 775347
Synonym trans-2-Butenedioic acid diethyl ester; Fumaric acid diethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1621422

 
alfa aesar™
A1621422
100g Each for $43.41

AAA162140E

 
alfa aesar™
A162140E
2500g Each for $538.74

AAA1621436

 
alfa aesar™
A1621436
500g Each for $124.36
Description & Specifications

Specifications

Formula Weight 172.18
Chemical Name or Material Diethyl fumarate
TSCA TSCA
Assay 98%
Molecular Formula C8H12O4
MDL Number MFCD00064455
Synonym trans-2-Butenedioic acid diethyl ester; Fumaric acid diethyl ester
Health Hazard 3 GHS P Statement
P210-P280-P264-P301+P312-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 91°C (195°F)
Boiling Point 98° to 100°C (14mmHg)
Quantity 100g
CAS 623-91-6
Beilstein 775347
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.
RTECS Number EM5950000
Melting Point ≈0°C

Applications
Diethyl fumarate, is used as an intermediate for plasticizer, flavoring agents and surface coatings. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It is also used as a intermediate and solvent in organic synthesis.

Solubility
It is soluble in water, acetone, and chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, acids, bases, reducing agents.

Safety and Handling

Warning

GHS P Statement
P210-P280-P264-P301+P312-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.