Please login to your online account to display your discounted pricing

Diethyl Formamidomalonate 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Diethyl formamidomalonate
Name Note 97%
CAS 6326-44-9
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H13NO5
Linear Formula C2H5O2CCH(NHCHO)CO2C2H5
Formula Weight 203.19
MDL Number MFCD00003279
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC114370050

  SDS
thermo fisher scientific
AC114370050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 203.19
Physical Form Adhering Crystalline Powder
Quantity 5g
Color White
EINECS Number 228-692-1
Melting Point 51° to 56°C
Health Hazard 1 Warning
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Assay Percent Range 97%
Name Note 97%
Linear Formula C2H5O2CCH(NHCHO)CO2C2H5
Chemical Name or Material Diethyl formamidomalonate
MDL Number MFCD00003279
CAS 6326-44-9
Molecular Formula C8H13NO5
Safety and Handling

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.