Learn More
Diethyl formamidomalonate, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC114370050
Specifications
Diethyl formamidomalonate | |
Authentic | |
173.0°C to 176.0°C (11.0mmHg) | |
51.0°C to 56.0°C | |
97% | |
96% min. (GC) | |
C2H5O2CCH(NHCHO)CO2C2H5 | |
04,47 | |
PFLHGSJLYNJIOF-UHFFFAOYSA-N | |
diethyl 2-formamidopropanedioate | |
80617 | |
Adhering Crystalline Powder |
113°C | |
Glass bottle | |
White | |
5g | |
6326-44-9 | |
C8H13NO5 | |
MFCD00003279 | |
1,3-diethyl 2-formamidopropanedioate, diethyl 2-carbonylaminopropane-1,3-dioate, diethyl 2-formamidomalonate, diethyl 2-formylaminomalonate, diethyl formamidomalonate, diethyl formaminomalonate, diethyl2-formylaminomalonate, diethylformaminomalonate, formamidomalonic acid, diethyl ester, malonic acid, formamido-, diethyl ester | |
CCOC(=O)C(C(=O)OCC)NC=O | |
203.19 | |
203.19 | |
97% |
Chemical Identifiers
6326-44-9 | |
203.19 | |
PFLHGSJLYNJIOF-UHFFFAOYSA-N | |
80617 | |
CCOC(=O)C(C(=O)OCC)NC=O |
C8H13NO5 | |
MFCD00003279 | |
1,3-diethyl 2-formamidopropanedioate, diethyl 2-carbonylaminopropane-1,3-dioate, diethyl 2-formamidomalonate, diethyl 2-formylaminomalonate, diethyl formamidomalonate, diethyl formaminomalonate, diethyl2-formylaminomalonate, diethylformaminomalonate, formamidomalonic acid, diethyl ester, malonic acid, formamido-, diethyl ester | |
diethyl 2-formamidopropanedioate |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
EINECSNumber : 228-692-1