Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl formamidomalonate, 97%

$47.77 - $162.73

Chemical Identifiers

Chemical Name or Material Diethyl formamidomalonate
CAS 6326-44-9
Molecular Formula C8H13NO5
Formula Weight 203.19
MDL Number MFCD00003279
Beilstein 1788903
Synonym Formamidomalonic acid diethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0118906

  SDS
alfa aesar™
L0118906
5g Each for $47.77

AAL0118914

  SDS
alfa aesar™
L0118914
25g Each for $162.73
Description & Specifications

Specifications

Formula Weight 203.19
Chemical Name or Material Diethyl formamidomalonate
MDL Number MFCD00003279
CAS 6326-44-9
Molecular Formula C8H13NO5
Synonym Formamidomalonic acid diethyl ester
Beilstein 1788903
Recommended Storage 2° to 8°C
Quantity 5g
Boiling Point 173° to 176°C (11mmHg)
Melting Point 50° to 55°C
Assay 97%
Safety and Handling

2° to 8°C