missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl Fluoromalonate, TCI America™

Manufacturer:  TCI America F028325G

 View more versions of this product

Catalog No. F028325G



Specifications

Specifications

Diethyl Fluoromalonate
Colorless
685-88-1
MFCD00009139
diethyl fluoromalonate, diethyl 2-fluoromalonate, fluoromalonic acid diethyl ester, 1,3-diethyl 2-fluoropropanedioate, diethylfluoromalonate, 2-fluoro-malonic acid diethyl ester, diethyl2-fluoromalonate, diethyl fluoropropanedioate, propanedioic acid, fluoro-, diethyl ester, malonic acid, fluoro-, diethyl ester
CCOC(=O)C(C(=O)OCC)F
178.159
178.16
≥95.0% (GC)
122°C
25g
C7H11FO4
1760
GOWQBFVDZPZZFA-UHFFFAOYSA-N
diethyl 2-fluoropropanedioate
12702
Clear Liquid at 20°C

Chemical Identifiers

685-88-1
178.159
GOWQBFVDZPZZFA-UHFFFAOYSA-N
12702
CCOC(=O)C(C(=O)OCC)F
C7H11FO4
MFCD00009139
diethyl fluoromalonate, diethyl 2-fluoromalonate, fluoromalonic acid diethyl ester, 1,3-diethyl 2-fluoropropanedioate, diethylfluoromalonate, 2-fluoro-malonic acid diethyl ester, diethyl2-fluoromalonate, diethyl fluoropropanedioate, propanedioic acid, fluoro-, diethyl ester, malonic acid, fluoro-, diethyl ester
diethyl 2-fluoropropanedioate
Safety and Handling

Safety and Handling

RTECSNumber : OO1670000

TSCA : No

Documents