Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl ethylphosphonate, 98%

$169.32 - $169.32

Chemical Identifiers

Chemical Name or Material Diethyl ethylphosphonate
CAS 78-38-6
Molecular Formula C6H15O3P
Formula Weight 166.16
MDL Number MFCD00039888
UN Number UN3082
Beilstein 1756861
Synonym Diethyl ethanephosphonate; Ethylphosphonic acid diethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA110460B

 
alfa aesar™
A110460B
1000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1104614

 
alfa aesar™
A1104614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1104630

 
alfa aesar™
A1104630
250g Each for $169.32
Description & Specifications

Specifications

Formula Weight 166.16
Chemical Name or Material Diethyl ethylphosphonate
TSCA TSCA
Assay 98%
Molecular Formula C6H15O3P
MDL Number MFCD00039888
CAS 78-38-6
UN Number UN3082
Boiling Point 82° to 83°C (11mmHg)
Beilstein 1756861
Flash Point 105°C (221°F)
Synonym Diethyl ethanephosphonate; Ethylphosphonic acid diethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P264-P305+P351+P338-P301+P312-P337+P313-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
RTECS Number SZ7925000
Quantity 1000g

Applications
Diethyl ethylphosphonate is used for heavy-metal extraction and solvent separation. It is also used as a gasoline additive, antifoam agent, plasticizer and textile conditioner. Further, it serves as an antistatic agent, flame retardant and as a general reagent in phosphinoalkene and phosphinoalkyne synthesis.

Solubility
Miscible with alcohol, ether and organic solvents. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280-P264-P305+P351+P338-P301+P312-P337+P313-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.