Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl ethylphenylmalonate, 96%

$223.00 - $223.00

Chemical Identifiers

Chemical Name or Material Diethyl ethylphenylmalonate
CAS 76-67-5
Molecular Formula C15H20O4
Formula Weight 264.32
MDL Number MFCD00040759
Beilstein 668299
Synonym Ethylphenylmalonic acid diethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0799814

  SDS
alfa aesar™
L0799814
25g Each for $223.00
Description & Specifications

Specifications

Formula Weight 264.32
Chemical Name or Material Diethyl ethylphenylmalonate
CAS 76-67-5
TSCA TSCA
Assay 96%
Molecular Formula C15H20O4
MDL Number MFCD00040759
Boiling Point 185°C (15mmHg)
Flash Point >110°C (230°F)
Synonym Ethylphenylmalonic acid diethyl ester
Quantity 25g
Beilstein 668299