missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl Ethylmalonate, TCI America™

Manufacturer:  TCI America E013825ML

 View more versions of this product

Catalog No. E013825ML



Specifications

Specifications

Diethyl Ethylmalonate
Colorless
133-13-1
MFCD00009167
VQAZCUCWHIIFGE-UHFFFAOYSA-N
diethyl 2-ethylpropanedioate
8610
Clear Liquid at 20°C
211°C
25mL
C9H16O4
diethyl ethylmalonate, diethyl 2-ethylmalonate, propanedioic acid, ethyl-, diethyl ester, ethylmalonic acid diethyl ester, 1,3-diethyl 2-ethylpropanedioate, unii-69g92oyr3m, propanedioic acid, 2-ethyl-, 1,3-diethyl ester, malonic acid, ethyl-, diethyl ester, diethyl ethylpropanedioate, diethyl 2-ethylpropane-1,3-dioate
CCC(C(=O)OCC)C(=O)OCC
188.223
188.22
≥98.0% (GC)

Chemical Identifiers

133-13-1
188.223
VQAZCUCWHIIFGE-UHFFFAOYSA-N
8610
CCC(C(=O)OCC)C(=O)OCC
C9H16O4
MFCD00009167
diethyl ethylmalonate, diethyl 2-ethylmalonate, propanedioic acid, ethyl-, diethyl ester, ethylmalonic acid diethyl ester, 1,3-diethyl 2-ethylpropanedioate, unii-69g92oyr3m, propanedioic acid, 2-ethyl-, 1,3-diethyl ester, malonic acid, ethyl-, diethyl ester, diethyl ethylpropanedioate, diethyl 2-ethylpropane-1,3-dioate
diethyl 2-ethylpropanedioate
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0264

TSCA : Yes

Documents