Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl ethylmalonate, 99%

$113.76 - $113.76

Chemical Identifiers

Chemical Name or Material Diethyl ethylmalonate
CAS 133-13-1
Molecular Formula C9H16O4
Formula Weight 188.22
MDL Number MFCD00009167
Beilstein 607635
Synonym Ethylmalonic acid diethyl ester; Ethylpropanedioic acid diethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2407736

 
alfa aesar™
B2407736
500g Each for $113.76
Description & Specifications

Specifications

Formula Weight 188.22
Chemical Name or Material Diethyl ethylmalonate
TSCA TSCA
Assay 99%
Molecular Formula C9H16O4
MDL Number MFCD00009167
CAS 133-13-1
Beilstein 607635
Synonym Ethylmalonic acid diethyl ester; Ethylpropanedioic acid diethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 88°C (190°F)
Boiling Point 204° to 206°C
Melting Point 0°C
Quantity 500g
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.