Learn More
Diethyl Ethoxymethylenemalonate, 99+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 114345000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Diethyl ethoxymethylenemalonate | |
87-13-8 | |
279°C to 281°C | |
Authentic | |
Glass bottle | |
1.4610 to 1.463 | |
MFCD00009148 | |
03, 469 | |
1.08 | |
Solubility in water: insoluble | |
CCOC=C(C(=O)OCC)C(=O)OCC | |
216.23 | |
216.23 |
99+% | |
1.0800g/mL | |
144°C | |
99% min. (GC) | |
C10H16O5 | |
C2H5OCH=C(CO2C2H5)2 | |
500g | |
05,216 | |
diethyl ethoxymethylenemalonate, diethyl 2-ethoxymethylene malonate, ethoxymethylenemalonic acid diethyl ester, diethyl ethoxymethylene malonate, diethyl emme, propanedioic acid, ethoxymethylene-, diethyl ester, diethylethoxymethylenemalonate, 1,3-diethyl 2-ethoxymethylidene propanedioate, diethyl ethoxymethylidene propanedioate | |
LTMHNWPUDSTBKD-UHFFFAOYSA-N | |
diethyl 2-(ethoxymethylidene)propanedioate | |
6871 | |
99+% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 201-725-7
RTECSNumber : OO1100000
TSCA : TSCA
RUO â Research Use Only