Please login to your online account to display your discounted pricing

Diethyl Ether for Spectroscopy Uvasol™ EMD Millipore

For spectroscopy Uvasol™

$339.29 - $339.29

Chemical Identifiers

Chemical Name or Material Diethyl Ether
CAS 60-29-7
Molecular Formula C4H10O
Formula Weight 74.12
UN Number 1155
Synonym Ethyl ether, Ether, Et2O
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1009301000

 
emd millipore corporation
1009301000
Glass Bottle 1L Each for $339.29
Description & Specifications

Specifications

Boiling Point 34.6°C
Formula Weight 74.12
Melting Point −116.3°C
Quantity 1L
UN Number 1155
Health Hazard 1 H224: Extremely flammable liquid and vapour.
Vapor Pressure 587 hPa (20°C)
CAS 60-29-7
Color Colorless
Storage Information 15° to 25°C (59° to 77°F); Keep container tightly closed. Keep container in a cool, well-ventilated place. Keep away from heat and source of ignition.
Density 0.71g/cm3 at 20°C
Chemical Name or Material Diethyl Ether
Physical Form Liquid
Solubility Information Soluble in water: 69g/L at 20°C
Odor Charactristic
Packaging Glass Bottle
Health Hazard 2 H302: Harmful if swallowed.
Ignition Point 180°C (356°F)
Flash Point −40°C (−40°F)
Description The Uvasol™ solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The Uvasol™ solvents offer best UV transmittance.
Health Hazard 3 H336: May cause drowsiness or dizziness.
Synonym Ethyl ether, Ether, Et2O
Molecular Formula C4H10O
Safety and Handling

H224: Extremely flammable liquid and vapour.

H336: May cause drowsiness or dizziness.

H302: Harmful if swallowed.