missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl ether, anhydrous, ACS, 99% min, stab. with BHT, Alfa Aesar™

Manufacturer:  Alfa Aesar 1676762

 View more versions of this product

Catalog No. AA1676762



Specifications

Specifications

Diethyl ether
60-29-7
C4H10O
MFCD00011646
14,3806
RTZKZFJDLAIYFH-UHFFFAOYSA-N
ethoxyethane
3283
74.12
≤99%
0.713g/mL
1.3525
34.6°C
-116°C
Anhydrous, Stabilized with BHT
99% min.
(C2H5)2O
UN1155
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
CCOCC
74.123
CHEBI:35702
Liquid
ACS
Sweet
−45° (−49°F)
Colorless
6 x 500g

Chemical Identifiers

60-29-7
74.123
RTZKZFJDLAIYFH-UHFFFAOYSA-N
3283
ethoxyethane
C4H10O
MFCD00011646
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
CHEBI:35702
CCOCC
Safety and Handling

Safety and Handling

GHS H Statement
H224-H302-H336
Extremely flammable liquid and vapor.
Harmful if swallowed.
May cause drowsiness or dizziness.

GHS P Statement
P210-P261-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: DIETHYL ETHER

RTECSNumber : KI5775000

TSCA : TSCA

SDS Documents