missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl ether, anhydrous, ACS, 99% min, stab. with BHT, Alfa Aesar™

Manufacturer:  Alfa Aesar 1676718

 View more versions of this product

Catalog No. AA1676718



Specifications

Specifications

Diethyl Ether
60-29-7
≤99%
C4H10O
MFCD00011646
14,3806
CCOCC
74.123
CHEBI:35702
Liquid
For ACS analysis
ACS Grade
99% min.
(C2H5)2O
UN1155
RTZKZFJDLAIYFH-UHFFFAOYSA-N
ethoxyethane
3283
74.12
50g

Chemical Identifiers

60-29-7
74.123
RTZKZFJDLAIYFH-UHFFFAOYSA-N
CHEBI:35702
CCOCC
C4H10O
MFCD00011646
3283
ethoxyethane
Safety and Handling

Safety and Handling

GHS H Statement
H224-H302-H336

GHS P Statement
P210-P261-P280-P240-P241-P303+P361+P353-P304+P340-P301+P312-P405-P501a

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: DIETHYL ETHER

RTECSNumber : KI5775000

TSCA : TSCA

SDS Documents