missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl ether, anhydrous, ACS, 99% min, stab. with BHT, Alfa Aesar™

Manufacturer:  Alfa Aesar 1676736

 View more versions of this product

Catalog No. AA1676736



Specifications

Specifications

Diethyl ether, anhydrous
60-29-7
C4H10O
MFCD00011646
14,3806
RTZKZFJDLAIYFH-UHFFFAOYSA-N
ethoxyethane
3283
74.12
≤99%
0.713
For ACS analysis
99% min.
(C2H5)2O
UN1155
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
CCOCC
74.123
CHEBI:35702
Liquid
ACS
500g

Chemical Identifiers

60-29-7
74.123
RTZKZFJDLAIYFH-UHFFFAOYSA-N
3283
ethoxyethane
C4H10O
MFCD00011646
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
CHEBI:35702
CCOCC
Safety and Handling

Safety and Handling

GHS H Statement
H224-H302-H336

GHS P Statement
P210-P261-P280-P240-P241-P303+P361+P353-P304+P340-P301+P312-P405-P501a

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: DIETHYL ETHER

RTECSNumber : KI5775000

TSCA : TSCA

SDS Documents