Learn More
Diethyl dicarbonate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2275322
Description
Ribonuclease inhibitorDiethyl dicarbonate acts as an inhibitor of ryanodine binding to ryanodine/calcium(II) receptor channel. It is useful for specific inactivation of nucleases during isolation of undegraded polynucleotides. Further, it inhibits platelet-activating factor acetyl hydrolase. In addition to this, it is involved in the modification reagent for His and Tyr residues in proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsRibonuclease inhibitorDiethyl dicarbonate acts as an inhibitor of ryanodine binding to ryanodine/calcium(II) receptor channel. It is useful for specific inactivation of nucleases during isolation of undegraded polynucleotides. Further, it inhibits platelet-activating factor acetyl hydrolase. In addition to this, it is involved in the modification reagent for His and Tyr residues in proteins.
Solubility
Miscible with chloroform, ethanol, esters, ketones and hydrocarbons. Slightly miscible with water.
Notes
Store in cool place. Incompatible with strong oxidizing agents, strong reducing agents, strong acids, strong bases and ammonia.
Specifications
Diethyl dicarbonate | |
1.101 | |
69°C (156°F) | |
C6H10O5 | |
MFCD00009106 | |
637031 | |
14,7998 | |
Miscible with chloroform,ethanol,esters,ketones and hydrocarbons. Slightly miscible with water. | |
CCOC(=O)OC(=O)OCC | |
162.14 | |
CHEBI:59051 | |
97% |
1609-47-8 | |
93°C to 94°C (18 mmHg) | |
0.97 | |
1.398 | |
100 g | |
Moisture sensitive | |
diethyl pyrocarbonate, diethyl dicarbonate, depc, diethyl oxydiformate, baycovin, diethylpyrocarbonate, ethyl pyrocarbonate, dicarbonic acid, diethyl ester, dicarbonic acid diethyl ester, diethyl pyrocarbonic acid | |
FFYPMLJYZAEMQB-UHFFFAOYSA-N | |
ethoxycarbonyl ethyl carbonate | |
3051 | |
162.14 | |
Liquid |
Chemical Identifiers
1609-47-8 | |
162.14 | |
FFYPMLJYZAEMQB-UHFFFAOYSA-N | |
3051 | |
ethoxycarbonyl ethyl carbonate |
C6H10O5 | |
MFCD00009106 | |
diethyl pyrocarbonate, diethyl dicarbonate, depc, diethyl oxydiformate, baycovin, diethylpyrocarbonate, ethyl pyrocarbonate, dicarbonic acid, diethyl ester, dicarbonic acid diethyl ester, diethyl pyrocarbonic acid | |
CHEBI:59051 | |
CCOC(=O)OC(=O)OCC |
Safety and Handling
GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302-H315-H319-H332-H335
EINECSNumber : 216-542-8
RTECSNumber : LQ9350000
TSCA : Yes
storageNote1 : Moisture Sensitive
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only