Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl di-n-propylmalonate, 97%

$36.47 - $432.45

Chemical Identifiers

Chemical Name or Material Diethyl di-n-propylmalonate
CAS 6065-63-0
Molecular Formula C13H24O4
Formula Weight 244.33
MDL Number MFCD00026840
Synonym Di-n-propylmalonic acid diethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2506214

  SDS
alfa aesar™
B2506214
25g Each for $36.47

AAB2506222

  SDS
alfa aesar™
B2506222
100g Each for $114.51

AAB2506236

  SDS
alfa aesar™
B2506236
500g Each for $432.45
Description & Specifications

Specifications

Formula Weight 244.33
Chemical Name or Material Diethyl di-n-propylmalonate
Assay 97%
Molecular Formula C13H24O4
MDL Number MFCD00026840
CAS 6065-63-0
Synonym Di-n-propylmalonic acid diethyl ester
Quantity 25g