missing translation for 'onlineSavingsMsg'
Learn More

Diethyl di-n-butylmalonate, 98+%, Thermo Scientific Chemicals

$168.42 - $168.42

Chemical Identifiers

CAS 596-75-8
Molecular Formula C15H28O4
Molecular Weight (g/mol) 272.385
MDL Number MFCD00026842
InChI Key WHKKUUPZLWUOIW-UHFFFAOYSA-N
Synonym diethyl dibutylmalonate, diethyl 2,2-dibutylmalonate, propanedioic acid, dibutyl-, diethyl ester, diethyl di-n-butylmalonate, diethyldibutylmalonate, malonic acid, dibutyl-, diethyl ester, propanedioic acid, 2,2-dibutyl-, 1,3-diethyl ester, 1,3-diethyl 2,2-dibutylpropanedioate, diethyl=dibutylmalonate, acmc-209mf2
PubChem CID 69000
IUPAC Name diethyl 2,2-dibutylpropanedioate
SMILES CCCCC(CCCC)(C(=O)OCC)C(=O)OCC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0990214
View Documents
Thermo Scientific Chemicals
L0990214
25 g
Each for $168.42
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

596-75-8
272.385
WHKKUUPZLWUOIW-UHFFFAOYSA-N
69000
CCCCC(CCCC)(C(=O)OCC)C(=O)OCC
C15H28O4
MFCD00026842
diethyl dibutylmalonate, diethyl 2,2-dibutylmalonate, propanedioic acid, dibutyl-, diethyl ester, diethyl di-n-butylmalonate, diethyldibutylmalonate, malonic acid, dibutyl-, diethyl ester, propanedioic acid, 2,2-dibutyl-, 1,3-diethyl ester, 1,3-diethyl 2,2-dibutylpropanedioate, diethyl=dibutylmalonate, acmc-209mf2
diethyl 2,2-dibutylpropanedioate

Specifications

596-75-8
150°C (12 mmHg)
1.433
25 g
diethyl dibutylmalonate, diethyl 2,2-dibutylmalonate, propanedioic acid, dibutyl-, diethyl ester, diethyl di-n-butylmalonate, diethyldibutylmalonate, malonic acid, dibutyl-, diethyl ester, propanedioic acid, 2,2-dibutyl-, 1,3-diethyl ester, 1,3-diethyl 2,2-dibutylpropanedioate, diethyl=dibutylmalonate, acmc-209mf2
CCCCC(CCCC)(C(=O)OCC)C(=O)OCC
272.385
272.39
Diethyl di-n-butylmalonate
0.95
C15H28O4
MFCD00026842
518421
WHKKUUPZLWUOIW-UHFFFAOYSA-N
diethyl 2,2-dibutylpropanedioate
69000
≥98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 209-890-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only