Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl cinnamylphosphonate, 98%

$159.72 - $518.32

Chemical Identifiers

Chemical Name or Material Diethyl cinnamylphosphonate
CAS 17316-55-1
Molecular Formula C13H19O3P
Formula Weight 254.27
MDL Number MFCD00798606
Synonym Cinnamylphosphonic acid diethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2110814

  SDS
alfa aesar™
B2110814
25g Each for $159.72

AAB2110822

  SDS
alfa aesar™
B2110822
100g Each for $518.32
Description & Specifications

Specifications

Formula Weight 254.27
Chemical Name or Material Diethyl cinnamylphosphonate
Molecular Formula C13H19O3P
CAS 17316-55-1
Assay 98%
Quantity 25g
MDL Number MFCD00798606
Synonym Cinnamylphosphonic acid diethyl ester