missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl chlorophosphite, 90%, tech., Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 114251000

 View more versions of this product

Catalog No. AC114251000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Diethyl chlorophosphite
589-57-1
1.0900g/mL
<1°C
88% min. (Argentometry)
C4H10ClO2P
(C2H5O)2PCl
100 mL
1.09
diethyl chlorophosphite, ethyl phosphorochloridite, diethyl phosphorochloridite, chlorodiethoxyphosphine, phosphorochloridous acid, diethyl ester, unii-f414s3v8k6, diethyl chlorophosphonite, chloro diethoxy phosphane, diethychlorophosphite, ethyl chlorophosphite
TXHWYSOQHNMOOU-UHFFFAOYSA-N
chloro(diethoxy)phosphane
68530
90%
Liquid
tech., 90%
Colorless to Yellow
153.0°C to 155.0°C
Authentic
Glass bottle
1.4380 to 1.4420
MFCD00009074
01,248
15, 3144
Solubility in water: reacts
CCOP(OCC)Cl
156.55
156.55
Technical

Chemical Identifiers

589-57-1
156.55
TXHWYSOQHNMOOU-UHFFFAOYSA-N
68530
CCOP(OCC)Cl
C4H10ClO2P
MFCD00009074
diethyl chlorophosphite, ethyl phosphorochloridite, diethyl phosphorochloridite, chlorodiethoxyphosphine, phosphorochloridous acid, diethyl ester, unii-f414s3v8k6, diethyl chlorophosphonite, chloro diethoxy phosphane, diethychlorophosphite, ethyl chlorophosphite
chloro(diethoxy)phosphane
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-652-2

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only