Learn More
Diethyl chlorophosphite, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0991914
Description
Diethyl chlorophosphite acts as a versatile phosphorylating agent for ester and lactone enolates, amines and alkenols. It is used in the synthesis of organophosphates and are considered anticholinesterase agents through observation in studies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiethyl chlorophosphite acts as a versatile phosphorylating agent for ester and lactone enolates, amines and alkenols. It is used in the synthesis of organophosphates and are considered anticholinesterase agents through observation in studies.
Solubility
Soluble in DMSO and chloroform. Reacts with water.
Notes
Air & Moisture Sensitive. Store away from air and strong bases. Incompatible with water, bases, active metals and oxidizing agents.
Specifications
Diethyl chlorophosphite | |
1.082 | |
25°C (77°F) | |
C4H10ClO2P | |
MFCD00009074 | |
UN2920 | |
Air and moisture sensitive | |
diethyl chlorophosphite, ethyl phosphorochloridite, diethyl phosphorochloridite, chlorodiethoxyphosphine, phosphorochloridous acid, diethyl ester, unii-f414s3v8k6, diethyl chlorophosphonite, chloro diethoxy phosphane, diethychlorophosphite, ethyl chlorophosphite | |
TXHWYSOQHNMOOU-UHFFFAOYSA-N | |
chloro(diethoxy)phosphane | |
68530 | |
97% |
589-57-1 | |
153°C to 155°C | |
Acrid | |
1.4375 | |
25 g | |
1098392 | |
14,3841 | |
Soluble in DMSO and chloroform. Reacts with water. | |
CCOP(OCC)Cl | |
156.546 | |
156.55 |
Chemical Identifiers
589-57-1 | |
156.546 | |
TXHWYSOQHNMOOU-UHFFFAOYSA-N | |
68530 | |
CCOP(OCC)Cl |
C4H10ClO2P | |
MFCD00009074 | |
diethyl chlorophosphite, ethyl phosphorochloridite, diethyl phosphorochloridite, chlorodiethoxyphosphine, phosphorochloridous acid, diethyl ester, unii-f414s3v8k6, diethyl chlorophosphonite, chloro diethoxy phosphane, diethychlorophosphite, ethyl chlorophosphite | |
chloro(diethoxy)phosphane |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber : 209-652-2
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only