Learn More
Diethyl chlorophosphate, 95%, Thermo Scientific Chemicals
$178.72 - $178.72
Chemical Identifiers
CAS | 814-49-3 |
---|---|
Molecular Formula | C4H10ClO3P |
Molecular Weight (g/mol) | 172.55 |
MDL Number | MFCD00009075 |
InChI Key | LGTLXDJOAJDFLR-UHFFFAOYSA-N |
Synonym | diethyl chlorophosphate, diethyl chlorophosphonate, diethyl phosphorochloridate, diethyl phosphorochloride, diethoxyphosphoryl chloride, o,o-diethyl chlorophosphate, diethoxyphosphorus oxychloride, phosphorochloridic acid, diethyl ester, o,o-diethyl chloridophosphate, o,o-diethyl chlorophosphonate |
PubChem CID | 13139 |
IUPAC Name | 1-[chloro(ethoxy)phosphoryl]oxyethane |
SMILES | CCOP(=O)(OCC)Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC407241000
|
Thermo Scientific Chemicals
407241000 |
100 g | Glass bottle |
Each for $178.72
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
814-49-3 | |
172.55 | |
LGTLXDJOAJDFLR-UHFFFAOYSA-N | |
13139 | |
CCOP(=O)(OCC)Cl |
C4H10ClO3P | |
MFCD00009075 | |
diethyl chlorophosphate, diethyl chlorophosphonate, diethyl phosphorochloridate, diethyl phosphorochloride, diethoxyphosphoryl chloride, o,o-diethyl chlorophosphate, diethoxyphosphorus oxychloride, phosphorochloridic acid, diethyl ester, o,o-diethyl chloridophosphate, o,o-diethyl chlorophosphonate | |
1-[chloro(ethoxy)phosphoryl]oxyethane |
Specifications
814-49-3 | |
1.1900g/mL | |
Authentic | |
Glass bottle | |
1.4160 to 1.4180 | |
MFCD00009075 | |
01,332 | |
1.19 | |
Solubility in water: may decompose. Other solubilities: soluble in chloroform | |
CCOP(=O)(OCC)Cl | |
172.55 | |
172.55 | |
Liquid |
Colorless to Yellow | |
81.0°C (6.0 mmHg) | |
94% min. (GC) | |
C4H10ClO3P | |
(C2H5O)2POCl | |
100 g | |
01,248; 03,98; 04,152; 05,217; 06,192; 08,171 | |
diethyl chlorophosphate, diethyl chlorophosphonate, diethyl phosphorochloridate, diethyl phosphorochloride, diethoxyphosphoryl chloride, o,o-diethyl chlorophosphate, diethoxyphosphorus oxychloride, phosphorochloridic acid, diethyl ester, o,o-diethyl chloridophosphate, o,o-diethyl chlorophosphonate | |
LGTLXDJOAJDFLR-UHFFFAOYSA-N | |
1-[chloro(ethoxy)phosphoryl]oxyethane | |
13139 | |
95% | |
Diethyl chlorophosphate, 95% |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Fatal if inhaled.
Fatal if swallowed.
Fatal in contact with skin.
Contact with water liberates toxic gas.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 212-396-4
RTECSNumber : TD1400000
TSCA : TSCA
RUO – Research Use Only