missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl carbonate, 99%, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific 114220010

 View more versions of this product

Catalog No. AC114220010



Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Alkylation reagent
Specifications

Specifications

Diethyl carbonate
0.9750g/mL
-43.0°C
33°C
Authentic
(C2H5O)2CO
03,5
01,247; 02,129; 03,95; 05,213
1.3830 to 1.3850
1L
OIFBSDVPJOWBCH-UHFFFAOYSA-N
CCOC(=O)OCC
118.13
118.13
99%
0.2% max. (K.F.)
105-58-8
Undesignated
126.0°C to 128.0°C
98.5% min. (GC)
C5H10O3
MFCD00009107
Glass bottle
15,3835
carbonic acid diethyl ester, carbonic acid, diethyl ester, carbonic ether, diaethylcarbonat, diatol, diethylkarbonat, ethoxyformic anhydride, ethyl carbonate, ethyl carbonate eto 2co, eufin
Solubility in water: 18.8g/L
0.975
diethyl carbonate
7766
Liquid
0.827 mPa.s (20°C)

Chemical Identifiers

105-58-8
118.13
OIFBSDVPJOWBCH-UHFFFAOYSA-N
7766
CCOC(=O)OCC
C5H10O3
MFCD00009107
carbonic acid diethyl ester, carbonic acid, diethyl ester, carbonic ether, diaethylcarbonat, diatol, diethylkarbonat, ethoxyformic anhydride, ethyl carbonate, ethyl carbonate eto 2co, eufin
diethyl carbonate
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-311-1

SDS Documents
Product Certifications
Promotions

Promotions

RUO – Research Use Only