Learn More
Diethyl bromomalonate, 90+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1096518
Description
Diethyl bromomalonate is used in perfumes. It is also used to synthesize other compounds such as barbiturates, artificial flavorings, vitamin B1, and vitamin B6.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiethyl bromomalonate is used in perfumes. It is also used to synthesize other compounds such as barbiturates, artificial flavorings, vitamin B1, and vitamin B6.
Solubility
Does not mix well with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store at room temperature. Incompatible with strong oxidizing agents and strong bases.
Specifications
Diethyl bromomalonate | |
1.402 | |
111°C (231°F) | |
1.455 | |
50 g | |
509768 | |
Does not mix well with water. | |
CCOC(=O)C(C(=O)OCC)Br | |
239.065 | |
239.07 |
685-87-0 | |
233°C to 235°C | |
C7H11BrO4 | |
MFCD00009138 | |
UN3265 | |
diethyl bromomalonate, diethyl 2-bromomalonate, bromomalonic acid diethyl ester, ethyl bromomalonate, diethylbromomalonate, propanedioic acid, bromo-, diethyl ester, 1,3-diethyl 2-bromopropanedioate, diethyl bromopropanedioate, malonic acid, bromo-, diethyl ester, propanedioic acid, 2-bromo-, 1,3-diethyl ester | |
FNJVDWXUKLTFFL-UHFFFAOYSA-N | |
diethyl 2-bromopropanedioate | |
69637 | |
≥90% |
Chemical Identifiers
685-87-0 | |
239.065 | |
FNJVDWXUKLTFFL-UHFFFAOYSA-N | |
69637 | |
CCOC(=O)C(C(=O)OCC)Br |
C7H11BrO4 | |
MFCD00009138 | |
diethyl bromomalonate, diethyl 2-bromomalonate, bromomalonic acid diethyl ester, ethyl bromomalonate, diethylbromomalonate, propanedioic acid, bromo-, diethyl ester, 1,3-diethyl 2-bromopropanedioate, diethyl bromopropanedioate, malonic acid, bromo-, diethyl ester, propanedioic acid, 2-bromo-, 1,3-diethyl ester | |
diethyl 2-bromopropanedioate |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 211-683-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only