Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl bis(2-cyanoethyl)malonate, 98+%

Chemical Identifiers

Chemical Name or Material Diethyl bis(2-cyanoethyl)malonate
CAS 1444-05-9
Molecular Formula C13H18N2O4
Formula Weight 266.3
MDL Number MFCD00013831
UN Number UN3439
Beilstein 1800053
Synonym Bis(2-cyanoethyl)malonic acid diethyl ester; Bis(4,4-ethoxycarbonyl)pimelonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2327706

  SDS
alfa aesar™
B2327706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2327714

  SDS
alfa aesar™
B2327714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2327722

  SDS
alfa aesar™
B2327722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 266.3
Chemical Name or Material Diethyl bis(2-cyanoethyl)malonate
TSCA TSCA
Assay ≥98%
Molecular Formula C13H18N2O4
MDL Number MFCD00013831
CAS 1444-05-9
UN Number UN3439
Boiling Point 174° to 176°C (0.5mmHg)
Beilstein 1800053
Melting Point 60° to 63°C
Synonym Bis(2-cyanoethyl)malonic acid diethyl ester; Bis(4,4-ethoxycarbonyl)pimelonitrile
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H312-H332
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H312-H332
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.