missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diethyl benzylmalonate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2289014
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Diethyl benzylmalonate | |
1.069 | |
>110°C (230°F) | |
1.486 | |
25 g | |
diethyl benzylmalonate, diethyl 2-benzylmalonate, benzylmalonic acid diethyl ester, propanedioic acid, phenylmethyl-, diethyl ester, unii-40y0x0xn6p, malonic acid, benzyl-, diethyl ester, malonic acid, 2-benzyl-, diethyl ester, diethyl benzyl malonate, 1,3-diethyl 2-benzylpropanedioate, diethyl benzylpropanedioate | |
CCOC(=O)C(CC1=CC=CC=C1)C(=O)OCC | |
250.294 | |
250.29 |
607-81-8 | |
162°C to 163°C (10 mmHg) | |
C14H18O4 | |
MFCD00009166 | |
615793 | |
ICZLTZWATFXDLP-UHFFFAOYSA-N | |
diethyl 2-benzylpropanedioate | |
69090 | |
97% |
Chemical Identifiers
607-81-8 | |
250.294 | |
ICZLTZWATFXDLP-UHFFFAOYSA-N | |
69090 | |
CCOC(=O)C(CC1=CC=CC=C1)C(=O)OCC |
C14H18O4 | |
MFCD00009166 | |
diethyl benzylmalonate, diethyl 2-benzylmalonate, benzylmalonic acid diethyl ester, propanedioic acid, phenylmethyl-, diethyl ester, unii-40y0x0xn6p, malonic acid, benzyl-, diethyl ester, malonic acid, 2-benzyl-, diethyl ester, diethyl benzyl malonate, 1,3-diethyl 2-benzylpropanedioate, diethyl benzylpropanedioate | |
diethyl 2-benzylpropanedioate |
Safety and Handling
EINECSNumber : 210-142-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only