Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl benzamidomalonate, 98+%

$281.77 - $281.77

Chemical Identifiers

Chemical Name or Material Diethyl benzamidomalonate
CAS 16798-45-1
Molecular Formula C14H17NO5
Formula Weight 279.29
MDL Number MFCD00015160
Beilstein 2817007
Synonym Benzamidomalonic acid diethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0237114

  SDS
alfa aesar™
L0237114
25g Each for $281.77
Description & Specifications

Specifications

Formula Weight 279.29
Chemical Name or Material Diethyl benzamidomalonate
CAS 16798-45-1
MDL Number MFCD00015160
Synonym Benzamidomalonic acid diethyl ester
Quantity 25g
Melting Point 61° to 65°C
Beilstein 2817007
Assay ≥98%
Molecular Formula C14H17NO5