Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl azodicarboxylate, 97%

$137.77 - $137.77

Chemical Identifiers

Chemical Name or Material Diethyl azodicarboxylate
CAS 1972-28-7
Molecular Formula C6H10N2O4
Linear Formula CH3CH2O2C-N=N-CO2CH2CH3
Formula Weight 174.16
MDL Number MFCD00009103
UN Number UN2810
Beilstein 908662
Synonym Azodicarboxylic acid diethyl ester; DEAD
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1934814

  SDS
alfa aesar™
L1934814
25g Each for $137.77

AAL1934806

  SDS
alfa aesar™
L1934806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1934822

  SDS
alfa aesar™
L1934822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 174.16
Chemical Name or Material Diethyl azodicarboxylate
Assay 97%
Molecular Formula C6H10N2O4
Linear Formula CH3CH2O2C-N=N-CO2CH2CH3
MDL Number MFCD00009103
CAS 1972-28-7
UN Number UN2810
Boiling Point 106°C (13mmHg)
Beilstein 908662
Flash Point 85°C (185°F)
Synonym Azodicarboxylic acid diethyl ester; DEAD
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H315-H319-H335-H227
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Quantity 25g

Applications
Diethyl azodicarboxylate acts as a dienophile used in Diels-Alder reactions. It is used as a reagent in alfa-thiocyanation of enolizable ketones with ammonium thiocyanate and annulation of N-protected imines. It acts as a reactant for the preparation of immunostimulants alfa-Galactosylceramides, bisubstrate inhibitors with molecular recognition at the active site of catechol-O-methyltransferase and aza-beta-lactams via NHC-catalyzed [2 + 2] cycloaddition with ketenes. It finds application in Mitsunobu reaction and used in the synthesis of pharmaceuticals like zidovudine and procarbazine. The presence of azo group in it is a Michael acceptor and used for the conversion of beta-ketoesters to the corresponding hydrazine derivatives in presence of copper(II) catalyst. Further, it is an efficient dehydrogenating agent, which is involved in the preparation of aldehydes, disulfides from alcohols and thiols respectively.

Solubility
Miscible with dichloromethane, diethyl ether and toluene.

Notes
Store in a cool place. Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.

Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H315-H319-H335-H227
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

2° to 8°C