Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl aminomethylenemalonate, 98%

$45.81 - $133.80

Chemical Identifiers

Chemical Name or Material Diethyl aminomethylenemalonate
CAS 6296-99-7
Molecular Formula C8H13NO4
Formula Weight 187.2
MDL Number MFCD01075695
Beilstein 1708651
Synonym Aminomethylenemalonic acid diethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1551103

  SDS
alfa aesar™
L1551103
1g Each for $45.81

AAL1551106

  SDS
alfa aesar™
L1551106
5g Each for $133.80
Description & Specifications

Specifications

Formula Weight 187.2
CAS 6296-99-7
Synonym Aminomethylenemalonic acid diethyl ester
Chemical Name or Material Diethyl aminomethylenemalonate
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Assay 98%
Health Hazard 1 Warning
MDL Number MFCD01075695
Health Hazard 2 GHS H Statement
H315-H319-H317-H335
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Boiling Point 114°C (1mmHg)
Melting Point 60° to 65°C
Beilstein 1708651
Molecular Formula C8H13NO4
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H317-H335
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.