Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl allylphosphonate, 96%

$43.86 - $623.41

Chemical Identifiers

Chemical Name or Material Diethyl allylphosphonate
CAS 1067-87-4
Formula Weight 178.17
MDL Number MFCD00015134
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1036503

 
alfa aesar™
1036503
1g Each for $43.86

AA1036506

 
alfa aesar™
1036506
5g Each for $160.08

AA1036514

 
alfa aesar™
1036514
25g Each for $623.41
Description & Specifications

Specifications

Formula Weight 178.17
Chemical Name or Material Diethyl allylphosphonate
Refractive Index 1.4350
Density 1.0356g/mL
Boiling Point 63°C (2.5mmHg)
Physical Form Liquid
Assay 96%
Quantity 1g
Flash Point >61° (141°F)
MDL Number MFCD00015134
CAS 1067-87-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.