Please login to your online account to display your discounted pricing

Diethyl allylmalonate, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC113960050

 View more versions of this product

Catalog No. AC113960050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 200.23
Physical Form Liquid
Assay ≥96.0%
Quantity 5g
Color Clear colorless to slightly yellow
TSCA TSCA
EINECS Number 218-072-9
Beilstein 02, 776
Refractive Index 1.4300 to 1.4320 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00009155
Name Note 97%
Molecular Formula C10H16O4
Linear Formula C2H5O2CCH(CH2CH=CH2)CO2C2H5
Chemical Name or Material Diethyl allylmalonate
Assay Percent Range 97%
CAS 2049-80-1

Specifications

Chemical Name or Material Diethyl allylmalonate
Name Note 97%
CAS 2049-80-1
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H16O4
Linear Formula C2H5O2CCH(CH2CH=CH2)CO2C2H5
Formula Weight 200.23
MDL Number MFCD00009155
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents