missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl adipate, 99%

Manufacturer:  Alfa Aesar™ A1147636

 View more versions of this product

Catalog No. AAA1147636



Description

Description

Applications
Poly(vinylidene flouride) (PVF2) gels in diethyl adipate (DEA) with fibrillar morphology is presented. Gelation process obeys the three-dimensional percolation mechanism. Combustion and emission characteristics of a direct-injection diesel engine is fueled with diesel-diethyl adipate blends.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

Diethyl adipate
250°C to 252°C
500g
C10H18O4
MFCD00009215
14,162
VIZORQUEIQEFRT-UHFFFAOYSA-N
diethyl hexanedioate
8844
202.25
>110°C (230°F)
-20°C
141-28-6
CH3CH2O2C(CH2)4CO2CH2CH3
1780035
diethyl adipate, ethyl adipate, 1,6-diethyl hexanedioate, hexanedioic acid, diethyl ester, adipic acid, diethyl ester, adipic acid diethyl ester, hexanedioic acid, diethyl, diethylhexanedioic acid, diethylester kyseliny adipove, ethyl delta-carboethoxyvalerate
CCOC(=O)CCCCC(=O)OCC
202.25
CHEBI:34697
99%

Chemical Identifiers

141-28-6
202.25
VIZORQUEIQEFRT-UHFFFAOYSA-N
8844
diethyl hexanedioate
C10H18O4
MFCD00009215
diethyl adipate, ethyl adipate, 1,6-diethyl hexanedioate, hexanedioic acid, diethyl ester, adipic acid, diethyl ester, adipic acid diethyl ester, hexanedioic acid, diethyl, diethylhexanedioic acid, diethylester kyseliny adipove, ethyl delta-carboethoxyvalerate
CHEBI:34697
CCOC(=O)CCCCC(=O)OCC
Safety and Handling

Safety and Handling

RTECSNumber : AV1100000

TSCA : TSCA

SDS Documents