Learn More
Diethyl acetylenedicarboxylate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 296351000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Diethyl acetylenedicarboxylate | |
762-21-0 | |
Yellow | |
107.0°C to 110.0°C (11.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4410 to 1.4430 | |
100 g | |
01,244; 02,127 | |
diethyl acetylenedicarboxylate, 2-butynedioic acid, diethyl ester, diethyl 2-butynedioate, 1,4-diethyl but-2-ynedioate, acetylenedicarboxylic acid, diethyl ester, acetylenedicarboxylic acid diethyl ester, diethylacetylenedicarboxylate, bis-ethoxycarbonyl acetylene, diethyl acetylene dicarboxylate, di-ethyl acetylenedicarboxylate | |
STRNXFOUBFLVIN-UHFFFAOYSA-N | |
diethyl but-2-ynedioate | |
69803 | |
97% |
97% | |
1.0°C to 3.0°C | |
1.0600g/mL | |
94°C | |
96% min. (GC) | |
C8H10O4 | |
MFCD00009186 | |
02, 803 | |
1.06 | |
Solubility in water: .. Other solubilities: soluble in ethyl alcohol,ethyl ether,carbon tetr | |
CCOC(=O)C#CC(=O)OCC | |
170.17 | |
170.17 | |
Liquid |
Chemical Identifiers
762-21-0 | |
170.17 | |
STRNXFOUBFLVIN-UHFFFAOYSA-N | |
69803 | |
CCOC(=O)C#CC(=O)OCC |
C8H10O4 | |
MFCD00009186 | |
diethyl acetylenedicarboxylate, 2-butynedioic acid, diethyl ester, diethyl 2-butynedioate, 1,4-diethyl but-2-ynedioate, acetylenedicarboxylic acid, diethyl ester, acetylenedicarboxylic acid diethyl ester, diethylacetylenedicarboxylate, bis-ethoxycarbonyl acetylene, diethyl acetylene dicarboxylate, di-ethyl acetylenedicarboxylate | |
diethyl but-2-ynedioate |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 212-095-8
RUO – Research Use Only