missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl acetonylsuccinate, 95%

$54.32 - $783.07

Chemical Identifiers

CAS 1187-74-2
Molecular Formula C11H18O5
Molecular Weight (g/mol) 230.26
MDL Number MFCD00071579
InChI Key AAPVOVKOHNCXJA-UHFFFAOYSA-N
Synonym 1,4-diethyl 2-2-oxopropyl butanedioate, acetonyl succinic, acetonyl succinic acid diethyl ester, acetonylsuccinic aci, acetonylsuccinic acid diethyl ester, butanedioic acid, 2-oxopropyl-, diethyl ester, diethyl 2-2-oxopropyl succinate, diethyl 2-2-oxopropyl succite, diethyl 2-acetonylbutanedioate, diethyl acetonylsuccinate
PubChem CID 5463816
IUPAC Name diethyl 2-(2-oxopropyl)butanedioate
SMILES CCOC(=O)CC(CC(=O)C)C(=O)OCC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6454506
View Documents
Alfa Aesar™
H6454506
5g Each for $54.32
 
AAH6454514
View Documents
Alfa Aesar™
H6454514
25g Each for $213.42
 
AAH6454522
View Documents
Alfa Aesar™
H6454522
100g Each for $783.07
 
Specifications

Chemical Identifiers

1187-74-2
230.26
AAPVOVKOHNCXJA-UHFFFAOYSA-N
5463816
CCOC(=O)CC(CC(=O)C)C(=O)OCC
C11H18O5
MFCD00071579
1,4-diethyl 2-2-oxopropyl butanedioate, acetonyl succinic, acetonyl succinic acid diethyl ester, acetonylsuccinic aci, acetonylsuccinic acid diethyl ester, butanedioic acid, 2-oxopropyl-, diethyl ester, diethyl 2-2-oxopropyl succinate, diethyl 2-2-oxopropyl succite, diethyl 2-acetonylbutanedioate, diethyl acetonylsuccinate
diethyl 2-(2-oxopropyl)butanedioate

Specifications

1.074 g/mL
135°C (1.3mmHg)
5g
C11H18O5
1911511
AAPVOVKOHNCXJA-UHFFFAOYSA-N
diethyl 2-(2-oxopropyl)butanedioate
5463816
Liquid
Diethyl acetonylsuccinate
1.441
Colorless to Yellow
1187-74-2
MFCD00071579
1,4-diethyl 2-2-oxopropyl butanedioate, acetonyl succinic, acetonyl succinic acid diethyl ester, acetonylsuccinic aci, acetonylsuccinic acid diethyl ester, butanedioic acid, 2-oxopropyl-, diethyl ester, diethyl 2-2-oxopropyl succinate, diethyl 2-2-oxopropyl succite, diethyl 2-acetonylbutanedioate, diethyl acetonylsuccinate
CCOC(=O)CC(CC(=O)C)C(=O)OCC
230.26
230.26
95%
Safety and Handling

Safety and Handling

EINECSNumber : 413-860-0

TSCA : No

Recommended Storage : Ambient temperatures