missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diethyl acetonylsuccinate, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6454514
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Diethyl acetonylsuccinate | |
Colorless to Yellow | |
135°C (1.3 mmHg) | |
1.441 | |
25 g | |
diethyl 2-2-oxopropyl succinate, diethyl acetonylsuccinate, acetonylsuccinic acid diethyl ester, acetonyl succinic acid diethyl ester, butanedioic acid, 2-oxopropyl-, diethyl ester, 1,4-diethyl 2-2-oxopropyl butanedioate, acetonyl succinic, acetonylsuccinic acid, diethyl 2-acetonylbutanedioate, diethyl 2-2-oxopropyl succite | |
CCOC(=O)CC(CC(C)=O)C(=O)OCC | |
230.26 | |
230.26 | |
Liquid |
1187-74-2 | |
1.074 g/mL | |
C11H18O5 | |
MFCD00071579 | |
1911511 | |
AAPVOVKOHNCXJA-UHFFFAOYNA-N | |
diethyl 2-(2-oxopropyl)butanedioate | |
5463816 | |
95% |
Chemical Identifiers
1187-74-2 | |
230.26 | |
AAPVOVKOHNCXJA-UHFFFAOYNA-N | |
5463816 |
C11H18O5 | |
MFCD00071579 | |
diethyl 2-2-oxopropyl succinate, diethyl acetonylsuccinate, acetonylsuccinic acid diethyl ester, acetonyl succinic acid diethyl ester, butanedioic acid, 2-oxopropyl-, diethyl ester, 1,4-diethyl 2-2-oxopropyl butanedioate, acetonyl succinic, acetonylsuccinic acid, diethyl 2-acetonylbutanedioate, diethyl 2-2-oxopropyl succite | |
CCOC(=O)CC(CC(C)=O)C(=O)OCC |
Safety and Handling
EINECSNumber : 413-860-0
TSCA : No
Recommended Storage : Ambient temperatures