Learn More
Diethyl Acetamidomalonate, 99+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 113931000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Diethyl acetamidomalonate | |
1068-90-2 | |
Authentic | |
Plastic bottle | |
C2H5O2CCH(NHCOCH3)CO2C2H5 | |
100g | |
diethyl acetamidomalonate, diethyl 2-acetamidomalonate, diethyl acetaminomalonate, diethyl acetylaminomalonate, acetamidomalonic acid diethyl ester, 2-acetylaminomalonic acid diethyl ester, 1,3-diethyl 2-acetamidopropanedioate, diethyl acetylamino malonate, propanedioic acid, acetylamino-, diethyl ester, acetylamino propanedioic acid diethyl ester | |
ISOLMABRZPQKOV-UHFFFAOYSA-N | |
diethyl 2-acetamidopropanedioate | |
14041 | |
99+% |
99+% | |
185°C (20.0mmHg) | |
99% min. (GC) | |
C9H15NO5 | |
MFCD00009146 | |
04, II, 891 | |
Solubility in water: insoluble. Other solubilities: soluble in ethanol | |
CCOC(=O)C(C(=O)OCC)NC(=O)C | |
217.22 | |
217.22 |
Chemical Identifiers
1068-90-2 | |
217.22 | |
ISOLMABRZPQKOV-UHFFFAOYSA-N | |
14041 | |
CCOC(=O)C(C(=O)OCC)NC(=O)C |
C9H15NO5 | |
MFCD00009146 | |
diethyl acetamidomalonate, diethyl 2-acetamidomalonate, diethyl acetaminomalonate, diethyl acetylaminomalonate, acetamidomalonic acid diethyl ester, 2-acetylaminomalonic acid diethyl ester, 1,3-diethyl 2-acetamidopropanedioate, diethyl acetylamino malonate, propanedioic acid, acetylamino-, diethyl ester, acetylamino propanedioic acid diethyl ester | |
diethyl 2-acetamidopropanedioate |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 213-952-9
RTECSNumber : OO0360000
TSCA : TSCA
RUO â Research Use Only