Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl 5-hydantoylphosphonate, 96%

$47.32 - $162.73

Chemical Identifiers

Chemical Name or Material Diethyl 5-hydantoylphosphonate
CAS 95378-36-2
Molecular Formula C7H13N2O5P
Formula Weight 236.17
MDL Number MFCD00171308
Beilstein 4809527
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1334103

  SDS
alfa aesar™
L1334103
1g Each for $47.32

AAL1334106

  SDS
alfa aesar™
L1334106
5g Each for $162.73
Description & Specifications

Specifications

Formula Weight 236.17
Chemical Name or Material Diethyl 5-hydantoylphosphonate
CAS 95378-36-2
MDL Number MFCD00171308
Assay 96%
Molecular Formula C7H13N2O5P
Melting Point 159° to 161°C
Beilstein 4809527
Quantity 1g